accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N(S)(R)OHOHOO

CAS RN 111900-32-4

IUPAC Name: (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Synonyms: (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C7H11NO4
InChi Key: YFYNOWXBIBKGHB-FBCQKBJTSA-N
SMILES: N[C@]1(CC[C@H](C1)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results