accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111918-90-2

IUPAC Name: 3-(methoxydimethylsilyl)propyl prop-2-enoate
Synonyms: 3-(methoxydimethylsilyl)propyl prop-2-enoate
Molecular Weight (g/mol): 202.33
Molecular Formula: C9H18O3Si
InChi Key: ZCRUJAKCJLCJCP-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)CCCOC(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results