accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-16-3

IUPAC Name: dodecanoyl chloride
Synonyms: dodecanoyl chloride
Molecular Weight (g/mol): 218.77
Molecular Formula: C12H23ClO
InChi Key: NQGIJDNPUZEBRU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (1)
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (5)
  • (4)
  • (7)
  • (1)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)