accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 112-37-8

IUPAC Name: undecanoic acid
Synonyms: undecanoic acid undekansaeure UDA
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: ZDPHROOEEOARMN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(O)=O
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)