accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CO

CAS RN 112-45-8

IUPAC Name: undec-10-enal
Synonyms: undecylenal
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: OFHHDSQXFXLTKC-UHFFFAOYSA-N
SMILES: C=CCCCCCCCCC=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)