accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-47-0

IUPAC Name: decane-1,10-diol
Synonyms: 1,10-decanediol
Molecular Weight (g/mol): 174.28
Molecular Formula: C10H22O2
InChi Key: FOTKYAAJKYLFFN-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (4)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (4)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (4)
  • (3)
  • (4)
  • (3)
  • (2)