accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH

CAS RN 112-50-5

IUPAC Name: 2-[2-(2-ethoxyethoxy)ethoxy]ethan-1-ol
Synonyms: ethoxytriethylene glycol
Molecular Weight (g/mol): 178.23
Molecular Formula: C8H18O4
InChi Key: WFSMVVDJSNMRAR-UHFFFAOYSA-N
SMILES: CCOCCOCCOCCO
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results