accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOH

CAS RN 112-60-7

IUPAC Name: 2-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: tetraethylene glycol peg-4 carbitol, diethyl HI-dry
Molecular Weight (g/mol): 194.23
Molecular Formula: C8H18O5
InChi Key: UWHCKJMYHZGTIT-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (5)
  • (6)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (2)
  • (5)
  • (5)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)