accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 112-66-3

IUPAC Name: dodecyl acetate
Synonyms: lauryl acetate
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: VZWGRQBCURJOMT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results