accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-72-1

IUPAC Name: tetradecan-1-ol
Synonyms: myristyl alcohol C14 alcohol epa pesticide chemical code 001510 lanette wax KS C14-15 alcohols dytol R-52 lanette K loxanol V fatty alcohol(C14) alcohol(C14)
Molecular Weight (g/mol): 214.39
Molecular Formula: C14H30O
InChi Key: HLZKNKRTKFSKGZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCO
Boiling Point 
  • (2)
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)