accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-76-5

IUPAC Name: octadecanoyl chloride
Synonyms: octadecanoyl chloride
Molecular Weight (g/mol): 302.93
Molecular Formula: C18H35ClO
InChi Key: WTBAHSZERDXKKZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(Cl)=O
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)