accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-84-5

IUPAC Name: (13Z)-docos-13-enamide
Synonyms: erucamide 13-decosenamide (cis)
Molecular Weight (g/mol): 337.59
Molecular Formula: C22H43NO
InChi Key: UAUDZVJPLUQNMU-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results