accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(Z)

CAS RN 112-86-7

IUPAC Name: (13Z)-docos-13-enoic acid
Synonyms: erucic acid delta 13-cis-docosenoic acid prifrac 2990 cis-delta(13)-docosenoic acid docos-13c-enoic acid delta13:14-docosenoic acid erucasaeure delta13-cis-docosenoic acid hystrene 2290 cis-eruic acid
Molecular Weight (g/mol): 338.58
Molecular Formula: C22H42O2
InChi Key: DPUOLQHDNGRHBS-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)