accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1120-04-3

IUPAC Name: sodium octadecyl sulfate
Synonyms: sodium stearyl sulfate
Molecular Weight (g/mol): 372.54
Molecular Formula: C18H37NaO4S
InChi Key: NWZBFJYXRGSRGD-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCCCCCOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (4)
  • (3)
  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)