accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 1120-57-6

IUPAC Name: chlorocyclobutane
Synonyms: chlorocyclobutane
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: STJYMUBZVMSMBP-UHFFFAOYSA-N
SMILES: ClC1CCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)