accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 1120-72-5

IUPAC Name: 2-methylcyclopentan-1-one
Synonyms: cyclopentanone, 2-methyl-
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ZIXLDMFVRPABBX-UHFFFAOYNA-N
SMILES: CC1CCCC1=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)