accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112077-84-6

IUPAC Name: carbonic acid; hydrazine
Synonyms: carbonic acid; hydrazine
Molecular Weight (g/mol): 94.07
Molecular Formula: CH6N2O3
InChi Key: PTYMQUSHTAONGW-UHFFFAOYSA-N
SMILES: NN.OC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results