accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1121-07-9

IUPAC Name: 1-methylpyrrolidine-2,5-dione
Synonyms: N-methylsuccinimide
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: KYEACNNYFNZCST-UHFFFAOYSA-N
SMILES: CN1C(=O)CCC1=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)