accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1121-25-1

IUPAC Name: 2-methylpyridin-3-ol
Synonyms: 2-methylpyridin-3-ol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: AQSRRZGQRFFFGS-UHFFFAOYSA-N
SMILES: CC1=NC=CC=C1O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)