accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1121-55-7

H2CN

CAS RN 1121-55-7

IUPAC Name: 3-ethenylpyridine
Synonyms: pyridine, 3-ethenyl-
Molecular Weight (g/mol): 105.14
Molecular Formula: C7H7N
InChi Key: DPZYLEIWHTWHCU-UHFFFAOYSA-N
SMILES: C=CC1=CC=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results