accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1122-41-4

IUPAC Name: (2,4-dichlorophenyl)sulfanide
Synonyms: (2,4-dichlorophenyl)sulfanide
Molecular Weight (g/mol): 178.05
Molecular Formula: C6H3Cl2S
InChi Key: FGBVJFREPSJSNG-UHFFFAOYSA-M
SMILES: [S-]C1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)