accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN

CAS RN 1122-71-0

IUPAC Name: (6-methylpyridin-2-yl)methanol
Synonyms: (6-methylpyridin-2-yl)methanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: JLVBSBMJQUMAMW-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CO)=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)