accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1122-99-2

IUPAC Name: 2-cyclopentylacetyl chloride
Synonyms: 2-cyclopentylacetyl chloride
Molecular Weight (g/mol): 146.61
Molecular Formula: C7H11ClO
InChi Key: NILLIUYSJFTTRH-UHFFFAOYSA-N
SMILES: ClC(=O)CC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results