accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112211-88-8

IUPAC Name: 3-methyl-2-[(propan-2-yl)amino]butan-1-ol
Synonyms: 2-(isopropylamino)-3-methylbutan-1-ol
Molecular Weight (g/mol): 145.25
Molecular Formula: C8H19NO
InChi Key: ANZBKMZVBJDTEL-UHFFFAOYNA-N
SMILES: CC(C)NC(CO)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results