accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112211-92-4

IUPAC Name: 2-phenyl-2-[(propan-2-yl)amino]ethan-1-ol
Synonyms: 2-(isopropylamino)-2-phenylethanol
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: XFSCVCVPECOHBL-UHFFFAOYNA-N
SMILES: CC(C)NC(CO)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results