accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOSOOHOSOOOHOHOHSOOOHSOOHO

CAS RN 112269-92-8

IUPAC Name: 25,26,27,28-tetrahydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaene-5,11,17,23-tetrasulfonic acid
Synonyms: 25,26,27,28-tetrahydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaene-5,11,17,23-tetrasulfonic acid
Molecular Weight (g/mol): 744.72
Molecular Formula: C28H24O16S4
InChi Key: JFYBCAFLVNKHHG-UHFFFAOYSA-N
SMILES: OC1=C2CC3=CC(=CC(CC4=C(O)C(CC5=C(O)C(CC1=CC(=C2)S(O)(=O)=O)=CC(=C5)S(O)(=O)=O)=CC(=C4)S(O)(=O)=O)=C3O)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)