accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1123-25-7

IUPAC Name: 1-methylcyclohexane-1-carboxylate
Synonyms: 1-methylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 141.19
Molecular Formula: C8H13O2
InChi Key: REHQLKUNRPCYEW-UHFFFAOYSA-M
SMILES: CC1(CCCCC1)C([O-])=O
Boiling Point 
  • (1)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)