accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1123-54-2

IUPAC Name: 2H-[1,2,3]triazolo[4,5-d]pyrimidin-7-amine
Synonyms: 2H-[1,2,3]triazolo[4,5-d]pyrimidin-7-amine
Molecular Weight (g/mol): 136.12
Molecular Formula: C4H4N6
InChi Key: HRYKDUPGBWLLHO-UHFFFAOYSA-N
SMILES: NC1=NC=NC2=NNN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results