accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1123491-15-5

IUPAC Name: 5,7-dimethoxy-3-(pyridin-4-yl)quinoline dihydrochloride
Synonyms: 5,7-dimethoxy-3-(pyridin-4-yl)quinoline dihydrochloride
Molecular Weight (g/mol): 339.22
Molecular Formula: C16H16Cl2N2O2
InChi Key: YBBAOKYVJCNJIV-UHFFFAOYSA-N
SMILES: Cl.Cl.COC1=CC(OC)=C2C=C(C=NC2=C1)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results