accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1124-08-9

IUPAC Name: 1,4-diiodo-2,5-dimethylbenzene
Synonyms: 1,4-diiodo-2,5-dimethylbenzene
Molecular Weight (g/mol): 357.96
Molecular Formula: C8H8I2
InChi Key: WJYYUEQVECTILJ-UHFFFAOYSA-N
SMILES: CC1=CC(I)=C(C)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results