accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3NH3C

CAS RN 1124-11-4

IUPAC Name: tetramethylpyrazine
Synonyms: tetramethylpyrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: FINHMKGKINIASC-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(C)N=C1C
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)