accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONO

CAS RN 1124-33-0

IUPAC Name: 4-nitropyridin-1-ium-1-olate
Synonyms: 4-nitropyridine-1-oxide
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: RXKNNAKAVAHBNK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=[N+]([O-])C=C1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)