accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1124-39-6

IUPAC Name: 4-ethylbenzene-1,2-diol
Synonyms: 4-ethylbenzene-1,2-diol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: HFLGBNBLMBSXEM-UHFFFAOYSA-N
SMILES: CCC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)