accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 1124-64-7

IUPAC Name: 1-butylpyridin-1-ium
Synonyms: 1-butylpyridin-1-ium
Molecular Weight (g/mol): 136.22
Molecular Formula: C9H14N
InChi Key: REACWASHYHDPSQ-UHFFFAOYSA-N
SMILES: CCCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)