accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNH3CCH3

CAS RN 112440-49-0

IUPAC Name: 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile
Synonyms: 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile
Molecular Weight (g/mol): 165.25
Molecular Formula: C9H11NS
InChi Key: GVRRCQCZKOPGDZ-UHFFFAOYSA-N
SMILES: CC1=C(C)C(CC#N)=C(C)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results