accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112486-09-6

IUPAC Name: 7,7-dimethyl-7H-benzo[c]fluorene
Synonyms: 7,7-dimethylbenzo[c]fluorene
Molecular Weight (g/mol): 244.34
Molecular Formula: C19H16
InChi Key: SJVOYVUTRSZMKW-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C3C=CC=CC3=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results