accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 1125-80-0

IUPAC Name: 3-methylisoquinoline
Synonyms: isoquinolone-3-methyl
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: FVVXWRGARUACNW-UHFFFAOYSA-N
SMILES: CC1=NC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results