accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHONHH2N

CAS RN 112522-64-2

IUPAC Name: N-(2-aminophenyl)-4-acetamidobenzamide
Synonyms: tacedinaline
Molecular Weight (g/mol): 269.3
Molecular Formula: C15H15N3O2
InChi Key: VAZAPHZUAVEOMC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results