accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112539-09-0

IUPAC Name: 4-(2-oxopyrrolidin-1-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 259.7
Molecular Formula: C10H10ClNO3S
InChi Key: OUAVTZYGHGCCCB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N1CCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results