accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 1126-00-7

IUPAC Name: 1-phenyl-1H-pyrazole
Synonyms: 1-phenyl-1H-pyrazole
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: WITMXBRCQWOZPX-UHFFFAOYSA-N
SMILES: C1=CN(N=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (5)