accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1126-61-0

IUPAC Name: 4-(prop-2-en-1-yl)benzene-1,2-diol
Synonyms: 4-allylpyrocatechol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: FHEHIXJLCWUPCZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC=C)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results