accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNN

CAS RN 112642-69-0

IUPAC Name: 2-octyl-2H-1,2,3-benzotriazole
Synonyms: 2-octyl-1,2,3-benzotriazole
Molecular Weight (g/mol): 231.34
Molecular Formula: C14H21N3
InChi Key: WESNTVKWORZVQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCN1N=C2C=CC=CC2=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results