accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112665-43-7

IUPAC Name: 7-phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
Synonyms: seratrodast bronica
Molecular Weight (g/mol): 354.45
Molecular Formula: C22H26O4
InChi Key: ZBVKEHDGYSLCCC-UHFFFAOYNA-N
SMILES: CC1=C(C)C(=O)C(C(CCCCCC(O)=O)C2=CC=CC=C2)=C(C)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results