accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNH3COCH3

CAS RN 1126779-11-0

IUPAC Name: 3,5-dimethyl-1-(oxan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 3,5-dimethyl-1-(oxan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 306.21
Molecular Formula: C16H27BN2O3
InChi Key: ZNDYJQBCSOGOJQ-UHFFFAOYNA-N
SMILES: CC1=NN(C2CCCCO2)C(C)=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results