accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHCNHO

CAS RN 1127-45-3

IUPAC Name: 8-hydroxy-1,4-dihydro-1λ²-quinoline-1,1-diium-4-id-1-olate
Synonyms: 8-hydroxy-1,4-dihydro-1λ²-quinoline-1,1-diium-4-id-1-olate
Molecular Weight (g/mol): 162.17
Molecular Formula: C9H8NO2
InChi Key: SYNVWFMRDDDWBS-UHFFFAOYNA-N
SMILES: OC1=CC=CC2=C1[NH++]([O-])C=C[CH-]2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results