accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112780-67-3

IUPAC Name: bromo(3-fluoro-4-methoxyphenyl)magnesium
Synonyms: bromo(3-fluoro-4-methoxyphenyl)magnesium
Molecular Weight (g/mol): 229.33
Molecular Formula: C7H6BrFMgO
InChi Key: PYDHWJRNBFAYJS-UHFFFAOYSA-M
SMILES: COC1=CC=C([Mg]Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results