accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1128-00-3

IUPAC Name: ethyl 2-aminocyclohex-1-ene-1-carboxylate
Synonyms: ethyl 2-aminocyclohex-1-ene-1-carboxylate
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: JBZVWABPSHNPIK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)CCCC1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)