accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3O

CAS RN 1128-08-1

IUPAC Name: 3-methyl-2-pentylcyclopent-2-en-1-one
Synonyms: dihydrojasmone
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: YCIXWYOBMVNGTB-UHFFFAOYSA-N
SMILES: CCCCCC1=C(C)CCC1=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)