accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1128-23-0

IUPAC Name: (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
Synonyms: L-gulonolactone L-(+)-gulono-1,4-lactone reduced ascorbic acid L-gulono-1,4-lactone gamma-gulonolactone reduced ascorbate L-gulonic acid gamma-lactone L-gulono-gamma-lactone
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: SXZYCXMUPBBULW-SKNVOMKLSA-N
SMILES: OC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)