accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 112809-25-3

IUPAC Name: 4-[(1H-1,2,4-triazol-1-yl)methyl]benzonitrile
Synonyms: 4-(1,2,4-triazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol): 184.2
Molecular Formula: C10H8N4
InChi Key: HQLYWHSJALKYOV-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(CN2C=NC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results